Dibenzosuberone, 97%
Dibenzosuberone, 97%
Dibenzosuberone, 97%
Thermo Scientific Chemicals

Dibenzosuberone, 97%

CAS: 1210-35-1 | C15H12O | 208.26 g/mol
製品番号(カタログ番号) A18405.14
または、製品番号A18405-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1210-35-1
Assay (GC)≥96.0%
Appearance (Color)Clear pale yellow to yellow
FormLiquid
Identification (FTIR)Conforms
Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.

Solubility
Soluble in water (0.03g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. J.C. Netto-Ferreira.; D. Weir.; J.C. Scaiano. Effect of ring structure on the photoreactivity of benzophenone-like molecules: a laser photolysis study of 10,10-dimethylanthrone and dibenzosuberone. Journal of Photochemistry and Photobiology A: Chemistry. 1989, 48 (2-3), 345-352.
  2. Sonja Merkas.; Mladen Litvic.; Ivica Cepanec.; Vladimir Vinkovic. Synthesis of Novel, Potentially Biologically Active Dibenzosuberone Derivatives. Molecules. 2005, 10 (12), 1429-1437.
  3. Flash vacuum pyrolysis gives anthracene in quantitative yield: Rec. Trav. Chim., 101, 317 (1982). For a study, with discussion of the mechanism, of the FVP of substituted dibenzosuberones to the corresponding anthracenes, see: J. Org. Chem., 60, 8407 (1995).