1,2-Propanediol, 98+%
1,2-Propanediol, 98+%
1,2-Propanediol, 98+%
Thermo Scientific Chemicals

1,2-Propanediol, 98+%

CAS: 57-55-6 | C3H8O2 | 76.095 g/mol
製品番号(カタログ番号) A18406.0E
または、製品番号A18406-0E
価格(JPY)
-
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS57-55-6
IUPAC Namepropane-1,2-diol
Molecular FormulaC3H8O2
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILESCC(O)CO
さらに表示
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1%
さらに表示

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
RUO – Research Use Only

General References:

  1. Feng, Y.; Yin, H.; Wang, A.; Xue, W. Selectively catalytic oxidation of 1,2-propanediol to lactic, formic, and acetic acids over Ag nanoparticles under mild reaction conditions. J. Catal. 2015, 326, 26-37.
  2. Vasiliadou, E. S.; Yfanti, V. L.; Lemonidou, A. A. One-pot tandem processing of glycerol stream to 1,2-propanediol with methanol reforming as hydrogen donor reaction. Appl. Catal., B 2015, 163, 258-266.