2,4,5-Trimethoxybenzoic acid, 99%
2,4,5-Trimethoxybenzoic acid, 99%
2,4,5-Trimethoxybenzoic acid, 99%
Thermo Scientific Chemicals

2,4,5-Trimethoxybenzoic acid, 99%

CAS: 490-64-2 | C10H11O5 | 211.19 g/mol
製品番号(カタログ番号) A19324.18
または、製品番号A19324-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS490-64-2
FormPowder
Appearance (Color)White
Melting Point (clear melt)141.0-147.0?C
Assay (Silylated GC)≥98.5%
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only