1-Bromo-4-chloro-2-fluorobenzene, 98%
1-Bromo-4-chloro-2-fluorobenzene, 98%
1-Bromo-4-chloro-2-fluorobenzene, 98%
Thermo Scientific Chemicals

1-Bromo-4-chloro-2-fluorobenzene, 98%

CAS: 1996-29-8 | C6H3BrClF | 209.44 g/mol
製品番号(カタログ番号) A19570.06
または、製品番号A19570-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1996-29-8
IUPAC Name1-bromo-4-chloro-2-fluorobenzene
Molecular FormulaC6H3BrClF
InChI KeyFPNVMCMDWZNTEU-UHFFFAOYSA-N
SMILESFC1=CC(Cl)=CC=C1Br
さらに表示
FormLow melting solid or clear colorless liquid
Appearance (Color)White/colorless to pale yellow
Refractive Index1.5520-1.5560 @ 20?C (as melt)
Assay (GC)≥97.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only