Citronellyl acetate, 96%
Citronellyl acetate, 96%
Citronellyl acetate, 96%
Thermo Scientific Chemicals

Citronellyl acetate, 96%

CAS: 150-84-5 | C12H22O2 | 198.306 g/mol
製品番号(カタログ番号) A19872.14
または、製品番号A19872-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS150-84-5
IUPAC Name3,7-dimethyloct-6-en-1-yl acetate
Molecular FormulaC12H22O2
InChI KeyJOZKFWLRHCDGJA-UHFFFAOYNA-N
SMILESCC(CCOC(C)=O)CCC=C(C)C
さらに表示
Appearance (Color)Clear colorless
Assay (GC)≥95.0%
FormLiquid
Refractive Index1.4415-1.4475 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only