2-Phenylethyl acetate, 98%
2-Phenylethyl acetate, 98%
2-Phenylethyl acetate, 98%
Thermo Scientific Chemicals

2-Phenylethyl acetate, 98%

CAS: 103-45-7 | C10H12O2 | 164.20 g/mol
製品番号(カタログ番号) B21238.30
または、製品番号B21238-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS103-45-7
IUPAC Name2-phenylethyl acetate
Molecular FormulaC10H12O2
InChI KeyMDHYEMXUFSJLGV-UHFFFAOYSA-N
SMILESCC(=O)OCCC1=CC=CC=C1
さらに表示
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4960-1.5000 @ 20?C
Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.

Solubility
It is insoluble in paraffin oil, water. Soluble in alcohol, dipropylene glycol, fixed oils, kerosene, propylene glycol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Protected from heat and light.
RUO – Research Use Only

General References:

  1. CE Fabre.; PJ Blanc.; G Goma. Production of 2-Phenylethyl Alcohol by Kluyveromyces marxianus. Biotechnology progress. 199814 (2), 270-274.
  2. JA Dickinson.; MR Hockridge.; RT Kroemer. Conformational Choice, Hydrogen Bonding, and Rotation of the S1 ← S0Electronic Transition Moment in 2-Phenylethyl Alcohol, 2-Phenylethylamine, and Their Water Clusters. J. Am. Chem. Soc. 1998120 (11), 2622-2632.