CAPSO, 98%
CAPSO, 98%
CAPSO, 98%
Thermo Scientific Chemicals

CAPSO, 98%

CAS: 73463-39-5 | C9H19NO4S | 237.314 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) B21305.36
または、製品番号B21305-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS73463-39-5
IUPAC Name(2S)-3-(cyclohexylazaniumyl)-2-hydroxypropane-1-sulfonate
Molecular FormulaC9H19NO4S
InChI KeyINEWUCPYEUEQTN-VIFPVBQESA-N
SMILESO[C@@H](C[NH2+]C1CCCCC1)CS([O-])(=O)=O
さらに表示
Melting Point (clear melt)263-273?C
Appearance (Color)White
FormCrystalline powder or flake
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
CAPSO is used in various biological applications such as electrophoresis. Specific applications include buffers for protein separations and diagnostic reagent production. Also used as biological buffer in the following studies, to maintain pH 9 during water leach test for the analysis of high-carbon coal fly ash-soil mixtures, chemiluminescent in situ hybridization using soybean peroxidase-labeled peptide nucleic acid (PNA) probes, preparation of planar bilayer lipid membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CAPSO is used in various biological applications such as electrophoresis. Specific applications include buffers for protein separations and diagnostic reagent production. Also used as biological buffer in the following studies, to maintain pH 9 during water leach test for the analysis of high-carbon coal fly ash-soil mixtures, chemiluminescent in situ hybridization using soybean peroxidase-labeled peptide nucleic acid (PNA) probes, preparation of planar bilayer lipid membranes.

Solubility
Soluble in water (10 g+ 90 ml).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Becker J, et al. Evaluation of leaching protocols for testing of high-carbon coal fly ash-soil mixtures. J. Environ. Eng., 2012, 139(5), 642-53.
  2. H Stender et. al. Rapid detection, identification, and enumeration of Escherichia coli cells in municipal water by chemiluminescent in situ hybridization. Applied and Environmental Microbiology, 2001, 67(1), 142-147.