(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride, 97%
(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride, 97%
(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride, 97%
Thermo Scientific Chemicals

(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride, 97%

CAS: 1253-47-0 | C27H24ClO2P | 446.91 g/mol
製品番号(カタログ番号) B21729.14
または、製品番号B21729-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1253-47-0
IUPAC Name{[4-(methoxycarbonyl)phenyl]methyl}triphenylphosphanium chloride
Molecular FormulaC27H24ClO2P
InChI KeyNKKAHXNONUTMEY-UHFFFAOYSA-M
SMILES[Cl-].COC(=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
さらに表示
FormCrystals or needles or powder or crystalline powder
Water Content (Karl Fischer Titration)< 3.0%
Appearance (Color)White to pale blue or pale grey
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only