Perfluoro-tert-butanol, 99%
Perfluoro-tert-butanol, 99%
Perfluoro-tert-butanol, 99%
Thermo Scientific Chemicals

Perfluoro-tert-butanol, 99%

CAS: 2378-02-1 | C4HF9O | 236.04 g/mol
製品番号(カタログ番号) B22442.03
または、製品番号B22442-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2378-02-1
IUPAC Name1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol
Molecular FormulaC4HF9O
InChI KeyXZNOAVNRSFURIR-UHFFFAOYSA-N
SMILESOC(C(F)(F)F)(C(F)(F)F)C(F)(F)F
さらに表示
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index<1.3000 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only