Diethyl dicarbonate, 97%
Diethyl dicarbonate, 97%
Diethyl dicarbonate, 97%
Thermo Scientific Chemicals

Diethyl dicarbonate, 97%

CAS: 1609-47-8 | C6H10O5 | 162.14 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) B22753.14
または、製品番号B22753-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1609-47-8
IUPAC Namediethyl dicarbonate
Molecular FormulaC6H10O5
InChI KeyFFYPMLJYZAEMQB-UHFFFAOYSA-N
SMILESCCOC(=O)OC(=O)OCC
さらに表示
Acid-base back titration≥96.0 to ≤104.0% (non-U.S. specification)
Assay from Suppliers CofA≥96.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms [non-U.S. specification]
Residue on ignition≤0.1% (U.S. specification)
さらに表示
Ribonuclease inhibitorDiethyl dicarbonate acts as an inhibitor of ryanodine binding to ryanodine/calcium(II) receptor channel. It is useful for specific inactivation of nucleases during isolation of undegraded polynucleotides. Further, it inhibits platelet-activating factor acetyl hydrolase. In addition to this, it is involved in the modification reagent for His and Tyr residues in proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ribonuclease inhibitorDiethyl dicarbonate acts as an inhibitor of ryanodine binding to ryanodine/calcium(II) receptor channel. It is useful for specific inactivation of nucleases during isolation of undegraded polynucleotides. Further, it inhibits platelet-activating factor acetyl hydrolase. In addition to this, it is involved in the modification reagent for His and Tyr residues in proteins.

Solubility
Miscible with chloroform, ethanol, esters, ketones and hydrocarbons. Slightly miscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids, strong bases and ammonia.
RUO – Research Use Only

General References:

  1. In the presence of the hindered base lithium dicyclohexylamide (from Dicyclohexyl amine, A15671), ɑ -ethoxycarbonylation of ketones occurs to give ß -keto esters: Synthesis, 1014 (1984).
  2. Dogandzhiyski, P.; Ghidini, A.; Danneberg, F.; Strömberg, R.; Göbel, M. W. Studies on Tris(2-aminobenzimidazole)-PNA Based Artificial Nucleases: A Comparison of Two Analytical Techniques. Bioconjugate Chem. 2015, 26 (12), 2514-2519.
  3. Zuldesmi, M.; Waki, A.; Kuroda, K.; Okido, M. Hydrothermal treatment of titanium alloys for the enhancement of osteoconductivity. Mater. Sci. Eng., C 2015, 49, 430-435.