2-(Dimethylamino)ethanol, 99+%
2-(Dimethylamino)ethanol, 99+%
2-(Dimethylamino)ethanol, 99+%
Thermo Scientific Chemicals

2-(Dimethylamino)ethanol, 99+%

CAS: 108-01-0 | C4H11NO | 89.138 g/mol
数量:
250 mL
1000 mL
製品番号(カタログ番号) B23616.AK
または、製品番号B23616-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS108-01-0
IUPAC Name2-(dimethylamino)ethan-1-ol
Molecular FormulaC4H11NO
InChI KeyUEEJHVSXFDXPFK-UHFFFAOYSA-N
SMILESCN(C)CCO
さらに表示
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥99.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤0.5%
さらに表示
2-(Dimethylamino)ethanol is used as corrosion inhibitor, anti-scaling agent, paint additive, coating additive and solids separation agent. It is also used as an intermediate for active pharmaceutical ingredients and dyes. It serves as a curing agent for polyurethanes and epoxy resins. Further, it is used as an additive to boiler water. In addition to this, it is used therapeutically as a CNS stimulant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water, alcohol, ether, acetone and benzene.

Notes
Incompatible with strong oxidizers and strong acids.
RUO – Research Use Only

General References:

  1. In the presence of 2 mol of n-BuLi, forms a unimetal superbase complex of the Li alkoxide with an additional molecule of n-BuLi: J. Chem. Soc., Perkin 1, 3071 (1997). For examples of its use, see 2-Methoxypyridine, A13675, 2-Chloropyridine, A14866.
  2. Hadri, N. E. L.; Quang, D. V.; Abu-Zahra, M. R. M. Study of Novel Solvent for CO2 Post-combustion Capture. Energy Procedia 2015, 75, 2268-2286.
  3. Bringas, G.; Navarro-Santos, P.; López-Rendón, R.; López-Lemus, J.; Bresme, F. Molecular Dynamics Simulations of 2-(Dimethylamino)ethanol. J. Phys. Chem. B 2015, 119 (15), 5035-5046.