4-Phenoxybenzonitrile, 96%
4-Phenoxybenzonitrile, 96%
4-Phenoxybenzonitrile, 96%
Thermo Scientific Chemicals

4-Phenoxybenzonitrile, 96%

CAS: 3096-81-9 | C13H9NO | 195.221 g/mol
製品番号(カタログ番号) B25364.04
または、製品番号B25364-04
価格(JPY)
-
数量:
2 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3096-81-9
IUPAC Name4-phenoxybenzonitrile
Molecular FormulaC13H9NO
InChI KeyUYHCIOZMFCLUDP-UHFFFAOYSA-N
SMILESN#CC1=CC=C(OC2=CC=CC=C2)C=C1
さらに表示
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥95.0%
Melting Point (clear melt)42.5-51.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only