Phosphate-Buffered Saline (PBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation.
This PBS is manufactured as follows:
Without
• Calcium
• Magnesium
• Phenol Red
Gibco® 'BASIC' products offer value-minded scientists liquid cell culture media and buffer solutions at a significant price savings. As part of Life Technologies’ commitment to quality, this select range of Research Use Only products is continually tested to ensure performance and customer satisfaction.
仕様
組成および保存条件
Storage conditions: 2–8°C. Protect from light. Shipping conditions: ambient Shelf life: 12 months from date of manufacture