Chlorobis[4-(trifluoromethyl)phenyl]phosphine, 97%
Chlorobis[4-(trifluoromethyl)phenyl]phosphine, 97%
Chlorobis[4-(trifluoromethyl)phenyl]phosphine, 97%
Thermo Scientific Chemicals

Chlorobis[4-(trifluoromethyl)phenyl]phosphine, 97%

CAS: 13685-24-0 | C14H8ClF6P | 356.632 g/mol
製品番号(カタログ番号) H25888.03
または、製品番号H25888-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13685-24-0
IUPAC Namechlorobis[4-(trifluoromethyl)phenyl]phosphane
Molecular FormulaC14H8ClF6P
InChI KeyPBLUOOURLNAOAJ-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(C=C1)P(Cl)C1=CC=C(C=C1)C(F)(F)F
さらに表示
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Proton NMRConforms to structure
Phosphorus-31 NMRConforms to structure
FormLiquid
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
RUO – Research Use Only
Hong-Yan Li,; Liang Zhou,; Ming-Yu Teng,; Qiu-Lei Xu,; Chen Lin,; You-Xuan Zheng,; Jing-Lin Zuo. Highly efficient green phosphorescent OLEDs based on a novel iridium complex. J. Mater. Chem. C. 2013, 1 (48), 560-565.