Agarose MS8, molecular sieve grade, for small DNA fragments
Agarose MS8, molecular sieve grade, for small DNA fragments
Agarose MS8, molecular sieve grade, for small DNA fragments
Thermo Scientific Chemicals

Agarose MS8, molecular sieve grade, for small DNA fragments

For small DNA fragments | CAS: 9012-36-6
製品番号(カタログ番号) H26738.14
または、製品番号H26738-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9012-36-6
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC24H38O19
InChI KeyMJQHZNBUODTQTK-WKGBVCLCSA-N
SMILESOC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
さらに表示
Appearance (Color)White to pale cream or pale yellow
Impurity contentSulfate, SO4: ≤0.25%
Residual water≤10%
FormPowder
Residue on ignition≤0.35%
Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. James M. Bailey.; Norman Davidson. Methylmercury as a reversible denaturing agent for agarose gel electrophoresis. Analytical Biochemistry. 1976, 70, (1), 75-85.
  2. Carl-Bertil Laurell. Quantitative estimation of proteins by electrophoresis in agarose gel containing antibodies. Analytical Biochemistry. 1966, 15, (1), 45-52.
  3. For molecular screening that improves resolution of small DNA fragments and polymerase chain reaction (PCR) products. Enhanced visibility and high gel strength.