Anthracene-9,10-diboronic acid bis(pinacol) ester, 95%
Anthracene-9,10-diboronic acid bis(pinacol) ester, 95%
Anthracene-9,10-diboronic acid bis(pinacol) ester, 95%
Thermo Scientific Chemicals

Anthracene-9,10-diboronic acid bis(pinacol) ester, 95%

CAS: 863992-56-7 | C26H32B2O4 | 430.16 g/mol
製品番号(カタログ番号) H51921.03
または、製品番号H51921-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS863992-56-7
IUPAC Name4,4,5,5-tetramethyl-2-[10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracen-9-yl]-1,3,2-dioxaborolane
Molecular FormulaC26H32B2O4
InChI KeyZLXSWNVYGSZXOP-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12
さらに表示
Assay (unspecified)94.0% min
It is used synthesis, structure and electrochemistry of mono- and dinuclear osmium n,n-di- and tetraphenylbipyridyls and extended bipyridyls. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used synthesis, structure and electrochemistry of mono- and dinuclear osmium n,n′-di- and tetraphenylbipyridyls and extended bipyridyls. It is also used as a primary and secondary intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Miguel A. Esteruelas, et al. Mono- and dinuclear osmium N,N'-di- and tetraphenylbipyridyls and extended bipyridyls. Synthesis, structure and electrochemistry.Dalton Trans.,2013,42(10), 3597-3608.