1,3-Dioxane, 98%
1,3-Dioxane, 98%
1,3-Dioxane, 98%
Thermo Scientific Chemicals

1,3-Dioxane, 98%

CAS: 505-22-6 | C4H8O2 | 88.106 g/mol
製品番号(カタログ番号) H54107.06
または、製品番号H54107-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS505-22-6
IUPAC Name1,3-dioxane
Molecular FormulaC4H8O2
InChI KeyVDFVNEFVBPFDSB-UHFFFAOYSA-N
SMILESC1COCOC1
さらに表示
FormLiquid
Purity≥97.5%
Appearance (Color)Colorless to almost colorless
1,3-Dioxane has been employed as solvent in the synthesis of 8- and 9-membered dioxazocines and dioxazonines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dioxane has been employed as solvent in the synthesis of 8- and 9-membered dioxazocines and dioxazonines.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Florian Medina; Céline Besnard; Jérôme Lacour. One-step synthesis of nitrogen-containing medium-sized rings via α-imino diazo intermediates. Organic Letters. 2015, 16 (12), 3232-3235.
  2. Fabio Marchetti; Guido Pampaloni; Stefano Zacchini. The reactivity of molybdenum pentachloride with ethers: routes to the synthesis of Mo(IV)Cl4 adducts, Mo(V) chlorido-alkoxides and Mo(V) oxydo-chlorides. Dalton Transactions (Print Edition). 2013, 42 (42), 15226-15234.