Potassium tetraphenylborate, 97%
Potassium tetraphenylborate, 97%
Potassium tetraphenylborate, 97%
Thermo Scientific Chemicals

Potassium tetraphenylborate, 97%

CAS: 3244-41-5 | C24H20BK | 358.332 g/mol
製品番号(カタログ番号) H55276.14
または、製品番号H55276-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3244-41-5
IUPAC Namepotassium tetraphenylboranuide
Molecular FormulaC24H20BK
InChI KeyMAMCUZQNCMYPCY-UHFFFAOYSA-N
SMILES[K+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to cream
FormPowder
It is an active pharmaceutical intemediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intemediate.

Solubility
Low solubility in water. Soluble in organic solvents.

Notes
Desiccate at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Joseph M. Mahoney.; Alicia M. Beatty.; Bradley D. Smith. Selective Recognition of an Alkali Halide Contact Ion-Pair. J. Am. Chem. Soc. 2001, 123 (24),.
  2. A. F. Findeis.; Thomas. de Vries. Polarography of Potassium Tetraphenylborate(III) in N,N-Dimethylformamide. Anal. Chem. 1956, 28 (2),.