(+)-Usnic acid, 98%
(+)-Usnic acid, 98%
(+)-Usnic acid, 98%
Thermo Scientific Chemicals

(+)-Usnic acid, 98%

CAS: 7562-61-0 | C18H16O7 | 344.32 g/mol
製品番号(カタログ番号) H56612.14
または、製品番号H56612-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7562-61-0
IUPAC Name(2S)-4,10-diacetyl-11,13-dihydroxy-2,12-dimethyl-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(13),6,9,11-tetraene-3,5-dione
Molecular FormulaC18H16O7
InChI KeyCUCUKLJLRRAKFN-KKIBXBACSA-N
SMILESCC(=O)C1C(=O)C=C2OC3=C(C(C)=O)C(O)=C(C)C(O)=C3[C@]2(C)C1=O
さらに表示
Assay (unspecified)>97.5%
It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary intermediate.

Solubility
Partly soluble in water. Soluble in acetone, ethanol, chloroform and furfural.

Notes
Store at 4°C. Reactive with oxidizing agents.
RUO – Research Use Only

General References:

  1. K. Ingólfsdóttir. Usnic acid. Phytochemistry. 2002, 61 (7),729-736.
  2. Moreno Cocchietto.; Nicola Skert.; Pier Nimis.; Gianni Sava. A review on usnic acid, an interesting natural compound. Naturwissenschaften. 2002, 89 (4),137-146.