1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as highly efficient privileged ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as highly efficient privileged ligands.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with oxidizing agents.
RUO – Research Use Only