Thermo Scientific Chemicals

4-Amino-5-bromo-2-hydroxypyrimidine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H61952.03
1 g, Each
Thermo Scientific Chemicals

4-Amino-5-bromo-2-hydroxypyrimidine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H61952.03
1 g, Each
数量
製品番号(カタログ番号): H61952.03
または、製品番号H61952-03

化学物質識別子

CAS
2240-25-7
IUPAC Name
6-amino-5-bromo-1,2-dihydropyrimidin-2-one
Molecular Formula
C4H4BrN3O
InChI Key
QFVKLKDEXOWFSL-UHFFFAOYSA-N
SMILES
NC1=C(Br)C=NC(=O)N1
Assay (HPLC)
>97.5%

概要

4-Amino-5-bromo-2-hydroxypyrimidine can be used in the synthesis of cross-link products under anaerobic and aerobic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-5-bromo-2-hydroxypyrimidine can be used in the synthesis of cross-link products under anaerobic and aerobic conditions.

Solubility
Soluble in formic acid (50 mg/ml).

Notes
Store away from st
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text