Thermo Scientific Chemicals

2-Bromo-1-fluoro-4-nitrobenzene, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号): H64045.14
25 g, Each
Thermo Scientific Chemicals

2-Bromo-1-fluoro-4-nitrobenzene, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号): H64045.14
25 g, Each
数量
製品番号(カタログ番号): H64045.14
または、製品番号H64045-14

化学物質識別子

CAS
701-45-1
IUPAC Name
2-bromo-1-fluoro-4-nitrobenzene
Molecular Formula
C6H3BrFNO2
InChI Key
FAWMTDSAMOCUAR-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(F)C(Br)=C1
Assay (HPLC)
>94.0%

概要

Reatant in the synthesis of N-fused tricyclic indoles, dimethyamine, benzofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of N-fused tricyclic indoles, dimethyamine, benzofuran.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text