Deoxyribonuclease I, bovine pancreas, Molecular Biology Grade
Thermo Scientific Chemicals

Deoxyribonuclease I, bovine pancreas, Molecular Biology Grade

製品番号(カタログ番号) J60027.EXU
または、製品番号J60027-!U
価格(JPY)
-
数量:
500 U
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9003-98-9
IUPAC NameDeoxyribonuclease
SMILES*
MDL NumberMFCD00130918
Unit definitionOne unit causes an increase in absorbance at 260nm of 0.001 per minute per ml, at 25?C and pH 5.0 when using highly polymerized DNA as a substrate
RNaseNone detected
Assay (unspecified)Units/mL: 2,000 min.
Protease activityNone detected
Endonuclease that splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide and removal of DNA from protein samples. Also used in is used to nick DNA, as a first step to incorporate labeled bases into DNA. The enzyme from Sigma has been used in the nuclease stock along with RNAse during cell lysate preparation of Madin-Darby canine kidney (MDCK) II cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only