Thermo Scientific Chemicals

Lactulose, 99%

製品番号(カタログ番号): J60160.22
100 g, Each
Thermo Scientific Chemicals

Lactulose, 99%

製品番号(カタログ番号): J60160.22
100 g, Each
数量
製品番号(カタログ番号): J60160.22
または、製品番号J60160-22

化学物質識別子

CAS
4618-18-2
IUPAC Name
(2S,3R,4S,5R,6R)-2-{[(2R,3S,4S,5R)-4,5-dihydroxy-2,5-bis(hydroxymethyl)oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C12H22O11
InChI Key
JCQLYHFGKNRPGE-FCVZTGTOSA-N
SMILES
OC[C@H]1O[C@](O)(CO)[C@@H](O)[C@@H]1O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Water Content (Karl Fischer Titration)
≤ 2%
Appearance (Color)
White
Assay (HPLC)
≥ 99%
Form
Powder

概要

It is not absorbed in the gut and is frequently used in the treatment of constipation and hepatic encephalopathy. actulose is also a food ingredient, better known as galactofructose, with sweet taste and offering beneficial health benefits on digestive health. Also used as a test of small intestine bacterial overgrowth (SIBO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (76.4%) at 30°C, water (81%) at 60°C, and water (>86%) at 90°C and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text