Methylene Blue trihydrate
Methylene Blue trihydrate
Methylene Blue trihydrate
Thermo Scientific Chemicals

Methylene Blue trihydrate

Sensitive stain for RNA and ribonuclease | CAS: 7220-79-3 | C16H24ClN3O3S | 373.90 g/mol
数量:
25 g
100 g
250 g
製品番号(カタログ番号) J60823.22
または、製品番号J60823-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7220-79-3
IUPAC Name3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium trihydrate chloride
Molecular FormulaC16H24ClN3O3S
InChI KeyXQAXGZLFSSPBMK-UHFFFAOYSA-M
SMILESO.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
さらに表示
Assay (unspecified)8 ppm Arsenic max.
Loss on Drying8.0 to 18.0%
Assay (unspecified)0.02% Zinc max.
Assay (unspecified)98-103%
Assay (unspecified)0.02% Copper max.
さらに表示
Methylene Blue trihydrate finds many uses in biology and chemistry. It acts as a GCS inhibitor, biological stain and redox indicator. It inhibits tau filament formation. Further, it is reported to bind to the M2 subtype of muscarinic receptors in rat cardiac myocytes and thus exhibit antimuscarinic effects on IK, ACh and ICa. It also inhibits endothelium-dependent relaxation of isolated blood vessels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylene Blue trihydrate finds many uses in biology and chemistry. It acts as a GCS inhibitor, biological stain and redox indicator. It inhibits tau filament formation. Further, it is reported to bind to the M2 subtype of muscarinic receptors in rat cardiac myocytes and thus exhibit antimuscarinic effects on IK, ACh and ICa. It also inhibits endothelium-dependent relaxation of isolated blood vessels.

Solubility
Soluble in water, alcohol and chloroform. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Xu, P.; Wang, J.; Xu, Y.; Chu, H.; Shen, H.; Zhang, D.; Zhao, M.; Liu, J.; Li, G. Binding Modes and Interaction Mechanism Between Different Base Pairs and Methylene Blue Trihydrate: A Quantum Mechanics Study. Adv. Struct. Bioinform. 2015, 827, 187-203.
  2. Chan, C. H.; Chia, C. H.; Zakaria, S.; Sajab, M. S.; Chin, S. X. Cellulose nanofibrils: a rapid adsorbent for the removal of methylene blue. RSC Adv. 2015, 5 (24), 18204-18212.