Transferrin (Apo), bovine plasma, 98+%
Thermo Scientific Chemicals

Transferrin (Apo), bovine plasma, 98+%

For research use
数量:
100mg
500mg
1g
製品番号(カタログ番号) J61626.03
または、製品番号J61626-03
価格(JPY)
-
数量:
1g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS11096-37-0
Assay (unspecified)(Beta Globulins 3% solution) Cellulose Acetate Electrophoresis: 95-100%
Assay (unspecified)Endotoxin by Limulus Amebocyte Lysate: 0-1.0 EU/mg
Water Content (Karl Fischer Titration)< 5.0%
pH7.0-8.0
Apo-Transferrin is also an important component used for the cultivation of mammalian cells in-vitro because it acts as a transport factor for defined culture media. This property is crucial for long-term mammalian cell growth in-vitro since ATF operates in media by binding to contaminating metal ions. Some other common uses for Apo-Transferrin are affinity purification and receptor mediated transfection of DNA-like molecules into cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only