Fast Blue BB base
Fast Blue BB base
Fast Blue BB base
Thermo Scientific Chemicals

Fast Blue BB base

CAS: 120-00-3 | C17H20N2O3 | 300.358 g/mol
製品番号(カタログ番号) J62456.14
または、製品番号J62456-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS120-00-3
IUPAC NameN-(4-amino-2,5-diethoxyphenyl)benzamide
Molecular FormulaC17H20N2O3
InChI KeyCNXZLZNEIYFZGU-UHFFFAOYSA-N
SMILESCCOC1=CC(NC(=O)C2=CC=CC=C2)=C(OCC)C=C1N
さらに表示
Assay (unspecified)Dye Content: 80% min.
Melting Point95-105?C
Assay (unspecified)UV, Lambda max: 318 ? 5 nm
C.I. 37175

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C.I. 37175
RUO – Research Use Only