Orange G, Electrophoresis Grade
Orange G, Electrophoresis Grade
Orange G, Electrophoresis Grade
Thermo Scientific Chemicals

Orange G, Electrophoresis Grade

Collagen stain for connective tissue | CAS: 1936-15-8 | C16H10N2Na2O7S2 | 452.36 g/mol
数量:
25 g
100 g
1 kg
製品番号(カタログ番号) J62743.14
または、製品番号J62743-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1936-15-8
IUPAC Namedisodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
Molecular FormulaC16H10N2Na2O7S2
InChI KeyAEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O
さらに表示
Appearance (Color)Orange to very dark orange and red to very dark red and orange-red and red-orange and dark red-orange
FormPowder
Assay (Dye)≥90%
Loss on Drying≤10.0%
lambda-max (UV-Vis)473-479nm (in water)
Collagen stain for connective tissue

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only