Vitamin A palmitate, 1.7 M.I.U./g
Vitamin A palmitate, 1.7 M.I.U./g
Vitamin A palmitate, 1.7 M.I.U./g
Thermo Scientific Chemicals

Vitamin A palmitate, 1.7 M.I.U./g

Retinyl palmitate is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. It is used as a dietary supplement as a lipophilic base for eg: soft gelatin capsule. | CAS: 79-81-2 | C36H60O2 | 524.874 g/mol
製品番号(カタログ番号) J63022.22
または、製品番号J63022-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS79-81-2
IUPAC Name(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl hexadecanoate
Molecular FormulaC36H60O2
InChI KeyVYGQUTWHTHXGQB-FFHKNEKCSA-N
SMILESCCCCCCCCCCCCCCCC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
さらに表示
Appearance (Color)Pale yellow to brown
FormLiquid or oil
Assay (unspecified)≥1.7 MIU/g
Assay from Supplier's CofA≥94.0% (HPLC)
Retinyl palmitate is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. It is used as a dietary supplement as a lipophilic base for eg: soft gelatin capsule. Other applications include as pharmaceutical product which is used for manufacturing aqueous solutions using solubilizing agents such as cremophor. It is also used in the fortification of fatty foods such as margarine, spreads, oils, milk and dairy products. It is used in cosmetics for the preparation of creams and lotions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only