Transfer or electro blotting buffer (10X), pH 8.5
Thermo Scientific Chemicals
Transfer or electro blotting buffer (10X), pH 8.5
Used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.
製品番号(カタログ番号) J63037.K7
または、製品番号J63037-K7
価格(JPY)
-
数量:
4L
一括またはカスタム形式をリクエストする
仕様
形状Liquid
数量4L
保存要件Ambient temperatures
Unit SizeEach
Transfer or electro blotting buffer (10X), pH 8.5 is used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
よくあるご質問(FAQ)
How can I prepare a 1X buffer solution from the 10X Transfer or electro blotting buffer (10X), pH 8.5?
You can make a 1:10 dilution of 10X Transfer or electro blotting buffer (10X), pH 8.5 with deionized water to create a 1X Transfer or electro blotting buffer. Adjust the pH to 8.5.
What are the concentrations of the components in the Transfer or electro blotting buffer (10X), pH 8.5?
Transfer or electro blotting buffer (10X), pH 8.5 consists of 250 mM Tris and 1.92 M glycine.