Trypsin, bovine pancreas
Trypsin, bovine pancreas
Trypsin, bovine pancreas
Thermo Scientific Chemicals

Trypsin, bovine pancreas

A pancreatic serine protease | CAS: 9002-07-7 | C35H47N7O10
製品番号(カタログ番号) J63688.09
または、製品番号J63688-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9002-07-7
IUPAC Nameacetic acid; tert-butyl N-(1-{[1-({4-[(diaminomethylidene)amino]-1-[(5-methyl-2-oxo-2H-chromen-7-yl)carbamoyl]butyl}carbamoyl)-2-hydroxyethyl]carbamoyl}-2-phenylethyl)carbamate
Molecular FormulaC35H47N7O10
InChI KeyWGWZNYKOUXOZTC-UHFFFAOYNA-N
SMILESCC(O)=O.CC1=C2C=CC(=O)OC2=CC(NC(=O)C(CCCN=C(N)N)NC(=O)C(CO)NC(=O)C(CC2=CC=CC=C2)NC(=O)OC(C)(C)C)=C1
さらに表示
FormLyophilized powder
Assay from Suppliers CofATrypsin Activity: ≥250 USP units/mg
Loss on Drying≤5.5%
Appearance (Color)White to off-white to cream
Residue on ignition≤2.5% (sulfated ash)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only
組成および保存条件
-30°C to -10°C