Albumin, Bovine, Fraction V, 96%, Standard Grade, New Zealand origin, pH 7.0
Thermo Scientific Chemicals

Albumin, Bovine, Fraction V, 96%, Standard Grade, New Zealand origin, pH 7.0

For research use
数量:
10 g
50 g
100 g
製品番号(カタログ番号) J65788.22
または、製品番号J65788-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9048-46-8
MDL NumberMFCD00145743
Heavy Metals (by ICP-OES)≤10 ppm
pH6.5-7.5 (10% solution in water)
Purity≥98.0%
Water Content≤5.0%
Appearance (Color)Off-white to yellow to green
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only