Polysorbate 20, Electrophoresis Reagent
Polysorbate 20, Electrophoresis Reagent
Polysorbate 20, Electrophoresis Reagent
Thermo Scientific Chemicals

Polysorbate 20, Electrophoresis Reagent

CAS: 9005-64-5 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | 522.68 g/mol
数量:
100 mL
500 mL
製品番号(カタログ番号) J66278.AE
または、製品番号J66278-AE
価格(JPY)
-
数量:
100 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9005-64-5
Molecular Formula(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
InChI KeyHMFKFHLTUCJZJO-UHFFFAOYNA-N
SMILESCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD00165986
Appearance (Color)Yellow
Assay (unspecified)Saponification Value: 40.00-50.00 KOH/g
FormLiquid
Water Content (Karl Fischer Titration)≤3.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only