Sodium glucoheptonate
Sodium glucoheptonate
Sodium glucoheptonate
Thermo Scientific Chemicals

Sodium glucoheptonate

CAS: 31138-65-5 | C7H13NaO8 | 248.163 g/mol
数量:
100 g
500 g
1 kg
製品番号(カタログ番号) J66811.36
または、製品番号J66811-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS31138-65-5
IUPAC Namesodium (3R,4S,5R,6R)-2,3,4,5,6,7-hexahydroxyheptanoate
Molecular FormulaC7H13NaO8
InChI KeyFMYOMWCQJXWGEN-BMZZJELJSA-M
SMILES[Na+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)C([O-])=O
さらに表示
Assay (unspecified)98.5% min.
Chloride content0.01% max.
Assay (unspecified)Sulfate: 0.1% max.
Water Content (Karl Fischer Titration)14.0% max.
Sodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.

Solubility
Freely soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Mehltretter, C. L.; Alexander, B. H.; Rist, C. E. Sequestration by Sugar Acids. Ind. Eng. Chem. 1953, 45, (12), 2782-2784.
  2. Kwamena E. Baidoo.; Susan Z. Lever. Synthesis of a diamine dithiol bifunctional chelating agent for incorporation of technetium-99m into biomolecules. Bioconjugate Chem. 1990, 1, (2), 132-137.