1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, 98%
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, 98%
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, 98%
Thermo Scientific Chemicals

1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, 98%

CAS: 25941-03-1 | C16H22O11 | 390.341 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) J66973.06
または、製品番号J66973-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS25941-03-1
IUPAC Name[3,4,5,6-tetrakis(acetyloxy)oxan-2-yl]methyl acetate
Molecular FormulaC16H22O11
InChI KeyLPTITAGPBXDDGR-UHFFFAOYNA-N
SMILESCC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
さらに表示
Assay (unspecified)Specific rotation: +51 to +57?C, c=1 in chloroform
Assay (unspecified)>97.5%
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is used for the preparation of glycopyranoside phosphates for the study of T-lymphocyte mediated inflammatory diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is used for the preparation of glycopyranoside phosphates for the study of T-lymphocyte mediated inflammatory diseases.

Solubility
Soluble in dichloromethane, ether, ethyl acetate, methanol, and chloroform.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store in cool place. Store at -20°C.
RUO – Research Use Only

General References:

  1. Qingbing Wang.; Jie Fu.; Jianbo Zhang. A facile preparation of peracylated α-aldopyranosyl chlorides with thionyl chloride and tin tetrachloride. Carbohydrate Research. 2008, 343, (17), 2989-2991.
  2. Sanjoy K Das.; M.Corazon Trono.; René Roy. Transition Metal-Catalyzed Cross-Coupling Reactions toward the Synthesis of α-d-Mannopyranoside Clusters. Methods in Enzymology. 2003, 362, 3-17.