Tris-HCl, 0.5M buffer soln., pH 8.0, low endotoxin
Tris-HCl, 0.5M buffer soln., pH 8.0, low endotoxin
Tris-HCl, 0.5M buffer soln., pH 8.0, low endotoxin
Thermo Scientific Chemicals

Tris-HCl, 0.5M buffer soln., pH 8.0, low endotoxin

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
数量:
100 mL
250 mL
製品番号(カタログ番号) J67510.AE
または、製品番号J67510-AE
価格(JPY)
-
数量:
100 mL
一括またはカスタム形式をリクエストする
仕様
CAS1185-53-1
化学物質名または材質TRIS-HCl
Colorless
IUPAC名2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
InChIキーQKNYBSVHEMOAJP-UHFFFAOYSA-N
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only