Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade
Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade
Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade
Thermo Scientific Chemicals

Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
数量:
100 g
250 g
500 g
製品番号(カタログ番号) J67606.30
または、製品番号J67606-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
さらに表示
pH6.0-7.5 (1% solution)
UV/VisAbsorbance 280 nm (3% in water): ≤0.4
Solution TestPasses (10% in water)
Assay from Suppliers CofA≥99.0% (unspecified)
Chloride content≤0.10%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only