Dichlorophenylsilane, 96%
Dichlorophenylsilane, 96%
Dichlorophenylsilane, 96%
Thermo Scientific Chemicals

Dichlorophenylsilane, 96%

CAS: 1631-84-1 | C6H5Cl2Si | 176.09 g/mol
製品番号(カタログ番号) L04371.14
または、製品番号L04371-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1631-84-1
IUPAC Namedichloro(phenyl)silyl
Molecular FormulaC6H5Cl2Si
InChI KeyXNAFLNBULDHNJS-UHFFFAOYSA-N
SMILESCl[Si](Cl)C1=CC=CC=C1
さらに表示
Assay (GC)≥95.0%
FormLiquid
Appearance (Color)Clear colorless
Dichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from moisture, oxidizing agents. Incompatible materials are bases, moisture and oxidizing agents.
RUO – Research Use Only
Atsutaka Kunai; Toshihiro Kawakami; Eiji Toyoda; Mitsuo Ishikawa. Highly selective synthesis of chlorosilanes from hydrosilanes.J. Am. Chem. Soc.1992, 11 (7), 2708-2711.