N-Benzoylaminopurine, 99%
N-Benzoylaminopurine, 99%
N-Benzoylaminopurine, 99%
Thermo Scientific Chemicals

N-Benzoylaminopurine, 99%

CAS: 4005-49-6 | C12H9N5O | 239.238 g/mol
製品番号(カタログ番号) L08292.14
または、製品番号L08292-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4005-49-6
IUPAC NameN-(7H-purin-6-yl)benzamide
Molecular FormulaC12H9N5O
InChI KeyQQJXZVKXNSFHRI-UHFFFAOYSA-N
SMILESO=C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1
さらに表示
FormCrystals or powder or crystalline powder or fused solid
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White to cream to pale brown
Assay (HPLC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only