1-Adamantaneacetic acid, 98+%
1-Adamantaneacetic acid, 98+%
1-Adamantaneacetic acid, 98+%
Thermo Scientific Chemicals

1-Adamantaneacetic acid, 98+%

CAS: 4942-47-6 | C12H17O2 | 193.27 g/mol
製品番号(カタログ番号) L12029.14
または、製品番号L12029-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4942-47-6
Melting Point (clear melt)133.0-139.0?C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.0%
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.

Solubility
Solubility in Methanol - almost transparency

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place
RUO – Research Use Only

General References:

  1. Małgorzata Śleszyńska.; Tomasz H. Wierzba.; Krzysztof Malinowski.; Tereza Tůmová.; Bernard Lammek.; Jiřina Slaninová.; Adam Prahl. Novel Bradykinin Analogues Modified in the N-Terminal Part of the Molecule with a Variety of Acyl Substituents. Int J Pept Res Ther. 2012, 18 (2), 117-124.
  2. Yong Chen.; Kun-Ying Han.; Yu Liu. Effective switch-on fluorescence sensing of zinc(II) ion by 8-aminoquinolino-β-cyclodextrin/adamantaneacetic acid system in water. Bioorganic & Medicinal Chemistry. 2007, 15 (3), 4537-4542.