Perfluorododecanoic acid, 96%, Thermo Scientific Chemicals
Perfluorododecanoic acid, 96%, Thermo Scientific Chemicals
Perfluorododecanoic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Perfluorododecanoic acid, 96%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
L14113.03
または、製品番号L14113-03
1 g
製品番号(カタログ番号) L14113.03
または、製品番号L14113-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS307-55-1
IUPAC Nametricosafluorododecanoic acid
Molecular FormulaC12HF23O2
InChI KeyCXGONMQFMIYUJR-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
さらに表示
Appearance (Color)White
FormPowder or crystals or flakes
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Identification (FTIR)Conforms
Melting Point (clear melt)102-117?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only