Triphenyltin chloride, 95%
Triphenyltin chloride, 95%
Triphenyltin chloride, 95%
Thermo Scientific Chemicals

Triphenyltin chloride, 95%

CAS: 639-58-7 | C18H17ClSn | 387.49 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) L14289.14
または、製品番号L14289-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS639-58-7
IUPAC Nametriphenyltin chloride
Molecular FormulaC18H17ClSn
InChI KeyZDYYCRIXXMWWQC-UHFFFAOYSA-N
SMILESCl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)White to pale cream
CommentAs a bulk commodity material may contain small quantites particulate matter from the manufacturing process
Melting Point (clear melt)103-109°C
Identification (FTIR)Conforms
FormCrystalline powder or powder
さらに表示
Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water (0.005g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to moisture.
RUO – Research Use Only

General References:

  1. L.E. Khoo.; J.-P. Charland, E.J. Gabe.; F.E. Smith. Synthesis and structural studies for a series of aromatic Schiff base complexes with triphenyltin chloride and triphenyltin isothiocyanate. Inorganica Chimica Acta. 1987, 128 (2), 139-145.
  2. N. G. Bokii.; G. N. Zakharova.; Yu. T. Struchkov. Crystal and molecular structures of triphenyltin chloride. Journal of Structural Chemistry. 1970, 11 (5), 828-835.