Trimellitic anhydride chloride, 98%
Trimellitic anhydride chloride, 98%
Trimellitic anhydride chloride, 98%
Thermo Scientific Chemicals

Trimellitic anhydride chloride, 98%

CAS: 1204-28-0 | C9H3ClO4 | 210.569 g/mol
製品番号(カタログ番号) L14382.14
または、製品番号L14382-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1204-28-0
IUPAC Name1,3-dioxo-1,3-dihydro-2-benzofuran-5-carbonyl chloride
Molecular FormulaC9H3ClO4
InChI KeyNJMOHBDCGXJLNJ-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=C2C(=O)OC(=O)C2=C1
さらに表示
Appearance (Color)White
FormCrystalline powder and/or lumps or fused solid (Monolithic or Coarse lumps) or flakes
Assay (GC)≥95.0% (non-U.S. specification)
Assay≤95.0 to ≤105.0% (Titration) (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only