2-Iodobenzyl alcohol, 98+%
2-Iodobenzyl alcohol, 98+%
2-Iodobenzyl alcohol, 98+%
Thermo Scientific Chemicals

2-Iodobenzyl alcohol, 98+%

CAS: 5159-41-1 | C7H7IO | 234.036 g/mol
製品番号(カタログ番号) L15126.14
または、製品番号L15126-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5159-41-1
IUPAC Name(2-iodophenyl)methanol
Molecular FormulaC7H7IO
InChI KeyWZCXOBMFBKSSFA-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1I
さらに表示
FormCrystals or powder or crystalline powder or needles
Melting Point (clear melt)88-93?C
Appearance (Color)White to yellow to pink or pale brown
Assay (GC)≥98.0%
2-Iodobenzyl alcohol was used in the synthesis of substituted seven-membered lactones, 2-[(E)-(1?-iodo-2?-propenyl)]benzyl alcohol2 and 2,3-diphenyl-1-indenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobenzyl alcohol was used in the synthesis of substituted seven-membered lactones, 2-[(E)-(1′-iodo-2′-propenyl)]benzyl alcohol2 and 2,3-diphenyl-1-indenone.

Solubility
Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Dinesh Kumar Rayabarapu; and Chien-Hong Cheng. Synthesis of seven-membered lactones via nickel- and zinc-catalyzed highly regio- and stereoselective cyclization of 2-iodobenzyl alcohols with propiolates. Journal of the American Chemical Society.2002, 124,(20), 5630-5631.
  2. Wang, G.; et al. Zirconium-catalyzed methylalumination of heterosubstituted arylethynes. Factors affecting the regio-, stereo-, and chemoselectivities. J. Organomet. Chem.2007, 692,4731.