Thermo Scientific Chemicals

1-(Fmoc-amino)cyclohexanecarboxylic acid, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): L19810.06
5 g, Each
Thermo Scientific Chemicals

1-(Fmoc-amino)cyclohexanecarboxylic acid, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): L19810.06
5 g, Each
数量
製品番号(カタログ番号): L19810.06
または、製品番号L19810-06

化学物質識別子

CAS
162648-54-6
Assay (Aqueous acid-base Titration)
≥98.0% to ≤102.0%
Form
Powder
Appearance (Color)
White
Assay (GC)
≥98.0%

概要

1-(Fmoc-amino)cyclohexanecarboxylic acid is an API so it is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals industry. Also used as raw materials for sodium fluosilicate, potassium fluosilicate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Fmoc-amino)cyclohexanecarboxylic acid is an API so it is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals industry. Also used as raw materials for sodium fluosilicate, potassium fluosilicate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text