Furan-3-boronic acid, 97%
Furan-3-boronic acid, 97%
Furan-3-boronic acid, 97%
Thermo Scientific Chemicals

Furan-3-boronic acid, 97%

CAS: 55552-70-0 | C4H5BO3 | 111.89 g/mol
製品番号(カタログ番号) L19834.06
または、製品番号L19834-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS55552-70-0
IUPAC Name(furan-3-yl)boronic acid
Molecular FormulaC4H5BO3
InChI KeyCYEFKCRAAGLNHW-UHFFFAOYSA-N
SMILESOB(O)C1=COC=C1
さらに表示
Appearance (Color)White to cream or grey
Assay (Aqueous acid-base Titration)≥96.0%
FormCrystals or powder or crystalline powder
Proton NMRConforms to structure
Assay (HPLC)≥96.0%
Furan-3-boronic acid use furan-3-boronic acid as our coupling partner in many reactions. A chitosan uricase (UOx)-poly (furan-3-boronic acid)(PFBA)-Pd nanoparticles (PdNPs)/plated Pd (Pd plate)/multiwalled carbon nanotubes (MWCNTs)/Au electrode was prepared for fabricating an amperometric biosensor and a biofuel cell (BFC) of uric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furan-3-boronic acid use furan-3-boronic acid as our coupling partner in many reactions. A chitosan uricase (UOx)-poly (furan-3-boronic acid)(PFBA)-Pd nanoparticles (PdNPs)/plated Pd (Pd plate)/multiwalled carbon nanotubes (MWCNTs)/Au electrode was prepared for fabricating an amperometric biosensor and a biofuel cell (BFC) of uric acid.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Christopher D. Spicera,; Benjamin G. Davis. Palladium-mediated site-selective Suzuki-Miyaura protein modification at genetically encoded aryl halides Chem. Commun. 2011, 47 issue ,1698-1700.
  2. Lingen Sun,; Jiali Liu,; Pei Zhang,; Yue Meng,; Chunhui Liu,; Yixuan Ma,; Qingji Xie. An amperometric biosensor and a biofuel cell of uric acid based on a chitosan/uricase-poly(furan-3-boronic acid)-Pd nanoparticles/plated Pd/multiwalled carbon nanotubes/Au electrode. Journal of Electroanalytical Chemistry. 2015 , 739 187-196.

よくあるご質問(FAQ)

How should I store Furan-3-boronic acid, 97%, Thermo Scientific Chemicals (Cat. No. L19834.06, L19834-06)?

We recommend storing Furan-3-boronic acid, 97%, Thermo Scientific Chemicals (Cat. No. L19834.06, L19834-06) in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.