6-Fluoropyridine-3-boronic acid, 95%
6-Fluoropyridine-3-boronic acid, 95%
6-Fluoropyridine-3-boronic acid, 95%
6-Fluoropyridine-3-boronic acid, 95%
Thermo Scientific Chemicals

6-Fluoropyridine-3-boronic acid, 95%

CAS: 351019-18-6 | C5H5BFNO2 | 140.91 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) L20387.06
または、製品番号L20387-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS351019-18-6
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥94.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only