Iodoform, 99%
Thermo Scientific™

Iodoform, 99%

High purity and versatile solvent suitable for HPLC and pesticide residue analysis | CAS: 540-84-1 | (CH3)3 CCH2CH(CH3)2 | 114.23 g/mol
製品番号(カタログ番号)数量パッケージ
O30144 LAmber Soda Lime Glass
製品番号(カタログ番号) O3014
価格(JPY)
-
数量:
4 L
パッケージ:
Amber Soda Lime Glass
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

High purity and versatile solvent suitable for HPLC and pesticide residue analysis

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only

Also available in recyclable FisherPak™ and NOWPak™ containers. EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

仕様
沸点98°C
CAS540-84-1
化学物質名または材質Isooctane
Colorless
融点-107°C
分子式(CH3)3 CCH2CH(CH3)2
パッケージAmber Soda Lime Glass
異名2,2,4-trimethylpentane
グレードHPLC/GC
分子量(g/mol)114.23
純度≥99%
数量4 L
Unit SizeEach