Search
Search

THY1| Gene Symbol: THY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001311160.2 | NP_001298089.1 | ||||
| NM_001311162.2 | NP_001298091.1 | |||||
| NM_001372050.1 | NP_001358979.1 | |||||
| NM_006288.5 | NP_006279.2 | |||||
| NR_164077.1 | ||||||
| GenBank mRNA | AB209209.1 | - | ||||
| AF261093.1 | - | |||||
| AK057865.1 | - | |||||
| AK090644.1 | - | |||||
| AK090996.1 | - | |||||
| AK094374.1 | - | |||||
| AK314018.1 | - | |||||
| AL161958.1 | - | |||||
| BC005175.2 | - | |||||
| BC037832.1 | - | |||||
| BC065559.1 | - | |||||
| BE672723.1 | - | |||||
| BM504140.1 | - | |||||
| DA063412.1 | - | |||||
| S59749.1 | - | |||||