Search
Search

IL12A| Gene Symbol: IL12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_000882.3 | NP_000873.2 | ||||
| GenBank mRNA | AF101062.1 | - | ||||
| AF180562.1 | - | |||||
| AW472810.1 | - | |||||
| BC035571.1 | - | |||||
| BC104982.1 | - | 8143985 | ||||
| BC104984.1 | - | 8143987 | ||||
| BG702253.1 | - | |||||
| ENST00000305579.0 | - | |||||
| M65271.1 | - | |||||
| M65291.1 | - | |||||